missing translation for 'onlineSavingsMsg'
Learn More

Allyl iodide, 97+%, stab. with copper, Thermo Scientific Chemicals

Catalog Number p-7081727
Click to view available options
Quantity:
50 g
250 g
556-56-9
C3H5I
167.98
MFCD00001094
HFEHLDPGIKPNKL-UHFFFAOYSA-N
allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene
11166
3-iodoprop-1-ene
ICC=C
556-56-9
C3H5I
167.98
MFCD00001094
HFEHLDPGIKPNKL-UHFFFAOYSA-N
allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene
11166
3-iodoprop-1-ene
ICC=C

Allyl iodide is used as a reagent used with allylindium sesquiiodide in the cis-double allylation of cyclopropenes. It is used in synthesis of other organic compounds such as N-alkyl 2-pyrrolidone, Sorbic acid ester etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl iodide is used as a reagent used with allylindium sesquiiodide in the cis-double allylation of cyclopropenes. It is used in synthesis of other organic compounds such as N-alkyl 2-pyrrolidone, Sorbic acid ester etc,.

Solubility
Soluble in alcohol, chloroform, ether, and diethyl ether. Insoluble in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents. Incompatible with heat, light, amines, nitrides, azo/diazo compounds, alkali metals, and epoxides.
TRUSTED_SUSTAINABILITY
Melting Point -99°C
Density 1.832
Boiling Point 101°C to 103°C
Flash Point 18°C (64°F)
Linear Formula CH2=CHCH2I
Refractive Index 1.554
Quantity 50 g
UN Number UN1723
Beilstein 1697594
Sensitivity Light sensitive
Merck Index 14,294
Solubility Information Soluble in alcohol,chloroform,ether,and diethyl ether. Insoluble in water.
Formula Weight 167.98
Percent Purity ≥97%
Chemical Name or Material Allyl iodide, Stabilized with copper
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'