missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl glycidyl ether, 99+%, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Melting Point | -100.0°C |
Color | Colorless |
Density | 0.9620g/mL |
Boiling Point | 154.0°C |
Flash Point | 48°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | AcroSeal™ Glass Bottle |
Show More |
Safety and Handling
Product Identifier
- Allyl glycidyl ether
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H361f-Suspected of damaging fertility.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'