missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allyl chloroformate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | Brown-Red to Yellow |
Density | 1.1300g/mL |
Boiling Point | 109.0°C to 110.0°C |
Flash Point | 31°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Linear Formula | ClCO2CH2CH=CH2 |
Show More |
Safety and Handling
Product Identifier
- Allyl chloroformate
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 3
- Acute toxicity Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'