missing translation for 'onlineSavingsMsg'
Learn More

Allyl chloroacetate, 98%, Thermo Scientific™

Catalog Number p-52031
Click to view available options
Quantity:
5 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
2916-14-5
C5H7ClO2
134.56
MFCD00058936
VMBJJCDVORDOCF-UHFFFAOYSA-N
allyl chloroacetate, allyl 2-chloroacetate, chloroacetic acid allyl ester, acetic acid, chloro-, 2-propenyl ester, prop-2-en-1-yl 2-chloroacetate, acetic acid, chloro-, allyl ester, allyl monochloroacetate, acmc-1cffa, chloroacetic acid,2-propenyl ester
76206
prop-2-enyl 2-chloroacetate
C=CCOC(=O)CCl
This item is not returnable. View return policy
2916-14-5
C5H7ClO2
134.56
MFCD00058936
VMBJJCDVORDOCF-UHFFFAOYSA-N
allyl chloroacetate, allyl 2-chloroacetate, chloroacetic acid allyl ester, acetic acid, chloro-, 2-propenyl ester, prop-2-en-1-yl 2-chloroacetate, acetic acid, chloro-, allyl ester, allyl monochloroacetate, acmc-1cffa, chloroacetic acid,2-propenyl ester
76206
prop-2-enyl 2-chloroacetate
C=CCOC(=O)CCl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1590g/mL
Boiling Point 164°C (265.0 mmHg)
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4450 to 1.447
Quantity 5 mL
Specific Gravity 1.159
Formula Weight 134.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Allyl chloroacetate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Allyl chloroacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'