missing translation for 'onlineSavingsMsg'
Learn More

Allyl(chloro)[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II), Thermo Scientific Chemicals

Catalog Number 15417185
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

The derivative of this compound serves as an exceptionally mild and versatile catalyst for Heck reactions of aryl chlorides and bromides. A sterically and electronically diverse array of aryl bromides, as well as activated aryl chlorides, couple with a range of mono- and disubstituted olefins at room temperature, furnishing the arylated product with high E/Z stereoselection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 1235509-04-2
Color Yellow
Molecular Formula C19H33ClNPPd
MDL Number MFCD25372543
Quantity 1 g
Synonym pdclallyl amphos, allyl chloro di-tert-butyl 4-dimethylaminophenyl phosphine palladium ii
Solubility Information Insoluble in water.
InChI Key OSTWSTKYPFRHAA-UHFFFAOYSA-M
SMILES CC(C)(C)P(C1=CC=C(C=C1)CNC)C(C)(C)C.[CH2-]C=C.Cl[Pd+]
IUPAC Name chloropalladium(1+);1-(4-ditert-butylphosphanylphenyl)-N-methylmethanamine;prop-1-ene
Molecular Weight (g/mol) 448.324
PubChem CID 73994976
Formula Weight 448.32
Physical Form Solid
Chemical Name or Material Allyl (chloro)[di-tert-butyl (4-dimethylaminophenyl)phosphine]palladium (II)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'