missing translation for 'onlineSavingsMsg'
Learn More

Alloxan monohydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-51959
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3237-50-1, 50-71-5
C4H4N2O5
MFCD00149399
ZIIHZVKHFWOENY-UHFFFAOYSA-N
This item is not returnable. View return policy
3237-50-1, 50-71-5
C4H4N2O5
MFCD00149399
ZIIHZVKHFWOENY-UHFFFAOYSA-N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Beilstein 24, 500
Merck Index 10, 276
Synonym isovaleraldehyde, 2,4-dinitrophenyl hydrazone, butanal, 3-methyl-, 2,4-dinitrophenyl hydrazone, isovaleraldehyde dnp, isovaleraldehyde, 2,4-dinitrophenylhydazone, 1e-3-methylbutanal 2,4-dinitrophenyl hydrazone #, isovaleraldehyde 2,4-dinitrophenyl hydrazone, butyraldehyde, 3-methyl-, 2,4-dinitrophenyl hydrazone, isovaleraldehyd-2,4-dinitrophenylhydrazone, isovaleraldehyde,4-dinitrophenyl hydrazone, n-e-3-methylbutylideneamino-2,4-dinitroaniline
SMILES OC1(O)C(=O)NC(=O)NC1=O
IUPAC Name N-[(E)-3-methylbutylideneamino]-2,4-dinitroaniline
Molecular Weight (g/mol) 160.09
PubChem CID 9569524
Formula Weight 160.09
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Alloxan monohydrate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'