missing translation for 'onlineSavingsMsg'
Learn More

Allantoin, 98%, Thermo Scientific Chemicals

Catalog Number p-51951
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
97-59-6
C4H6N4O3
158.12
MFCD00005260
POJWUDADGALRAB-UHFFFAOYSA-N
allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream
204
CHEBI:15676
(2,5-dioxoimidazolidin-4-yl)urea
C1(C(=O)NC(=O)N1)NC(=O)N
This item is not returnable. View return policy
97-59-6
C4H6N4O3
158.12
MFCD00005260
POJWUDADGALRAB-UHFFFAOYSA-N
allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream
204
CHEBI:15676
(2,5-dioxoimidazolidin-4-yl)urea
C1(C(=O)NC(=O)N1)NC(=O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 230.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 100 g
Beilstein 25, V,15, 338
Merck Index 15, 250
Solubility Information Solubility in water: 1g/190mL. Other solubilities: readily soluble in alkalies,1g/500mL alcohol,practically insoluble in ether
Formula Weight 158.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Allantoin, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'