missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase Mouse, Clone: ALPL/597, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | Alkaline Phosphatase |
---|---|
Clone | ALPL/597 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960048
|
EnquireBio
249-MSM1-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950048
|
EnquireBio
249-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Alkaline Phosphatase | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
249 | |
Recombinant human ALPL protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
55kDa |
ALPL/597 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_016856392 | |
ALPL | |
IgG1 κ | |
Protein A/G Purified | |
Cell Surface | |
Intestine |