missing translation for 'onlineSavingsMsg'
Learn More

Aliquat™ 336, average MW 442, ACROS Organics™

Brand:  Acros Organics 194970025

Code : 29

Additional Details : CAS Number : 63393-96-4 Weight : 3.70000kg Transport : UN number : 2922 Chem class : 1a Pack group : III


Quantity 2.5L
Packaging Glass bottle

 View more versions of this product

Product Code. 10542164

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Aliquat™ 336
63393-96-4
442
Average M.W. 442
88.2 to 90.6%, 112.0 to 115.0 (quaternary value)
MFCD00011862

Specifications

Aliquat™ 336
63393-96-4
Authentic
Viscous Liquid
1.4655 to 1.4715
MFCD00011862
5% max.
2.5L
Average M.W. 442
0.8800g/mL
Glass bottle
88.2 to 90.6%, 112.0 to 115.0 (quaternary value)
442
05,460; 06,404; 07,380; 13,192
Amber
132°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Aliquat]TS 336

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS