missing translation for 'onlineSavingsMsg'
Learn More

Alginic Acid, Sodium Salt, Low Viscosity, MP Biomedicals

Catalog Number p-3572862
Click to view available options
Quantity:
250 g
500 g
1 kg
This item is not returnable. View return policy
9005-38-3
(C6H7O7)A(C6H7O7)BNa
Sodium alginate
This item is not returnable. View return policy
9005-38-3
(C6H7O7)A(C6H7O7)BNa
Sodium alginate

Source: kelp

Color White to Cream
Physical Form Powder
Quantity 250 g
Assay Percent Range >98%
Linear Formula (C6H7O6Na)n
Chemical Name or Material Alginic Acid, Sodium Salt, Low Viscosity, Viscosity (1% aq. solution): <300cps
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'