Learn More
Alfa Aesar™ Yttria Stabilized Zirconium Oxide Conical Crucibles, Low Form
Brand:  Alfa Aesar™ 041286.KF
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 15423835
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
Zirconium Oxide
100 mL
55mm
50mm
1 Piece
71mm
Yttria Stabilized Zirconium Oxide Conical Crucibles, Low Form