Learn More
Thermo Scientific Chemicals Tammann Al-23 Crucibles
Brand:  Thermo Scientific™ 033060.KF
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15483545
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
280mm
1 Piece
20mm
Tammann Al-23 Crucibles
Al-23
90 mL
24mm