missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Magnesium Oxide Crucibles

Brand:  Alfa Aesar™ 042988.KF

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15433885

  • £264.60 / Each


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Specifications

20mm
Cylindrical
Magnesium Oxide
1 Piece
37mm
Magnesium Oxide Crucible
Videos
SDS