missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Glove Bag Clips
Click to view available options
Quantity:
36 Pk.
6 Pk.
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- For convenient hanging of glove storage bag
- Available in packs of 6 or 36
Specifications
Content And Storage | Ambient temperatures |
For Use With (Equipment) | Glove Storage Bags |
Quantity | 6 Pk. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'