Learn More
Thermo Scientific Chemicals Glassy Carbon Boats
Offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids
£184.00 - £776.00
Product Code | Brand | Dimensions (L x W x H) | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Capacity (Metric) | Price | Quantity & Availability | ||||
15406295
|
Thermo Scientific Chemicals
039026.KT |
29 x 16 x 10mm | 2 mL |
£184.00
Each |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15416295
|
Thermo Scientific Chemicals
039027.KT |
53 x 16 x 10mm | 3 mL |
£244.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15426295
|
Thermo Scientific Chemicals
039028.KT |
103 x 16 x 10mm | 10 mL |
£321.00
Each |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15486345
|
Thermo Scientific™
040999.KT |
76 x 33 x 25mm | 40 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Resistance to all wet decomposition agents and abrasive wear
- No memory effects or contamination of analytical samples
- Stability to acid and alkaline melts
- No wetting effect of metal melts
- Resistance to thermal shock allows rapid heating and cooling times
- Good electrical conductivity
- Used in vessels for ultra-high purity materials technology such as semiconductor connections and crystal growing such as doped halogenide crystals
RUO – Research Use Only