missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ALCAM/CD166 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
POA
Specifications
Antigen | ALCAM/CD166 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Adhesion Blockade |
Applications | Western Blot, Adhesion Blockade |
Classification | Polyclonal |
Description
ALCAM/CD166 Polyclonal specifically detects ALCAM/CD166 in Human samples. It is validated for Western Blot, Adhesion Blockade.Specifications
ALCAM/CD166 | |
Western Blot 0.1 ug/mL, Adhesion Blockade | |
Polyclonal | |
Goat | |
Human | |
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
ALCAM | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Adhesion Blockade | |
Unconjugated | |
RUO | |
AAB59499 | |
214 | |
Mouse myeloma cell line NS0-derived recombinant human ALCAM/CD166 Trp28-Ala526 Accession # AAB59499 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human ALCAM/CD166 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse ALCAM/CD166 is observed. |
For Research Use Only