missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Albumin Fraction V

Derived from bovine serum and packaged in glass bottles for use in biochemistry.

Brand:  Merck Millipore 1120180100

Code : NEW

Additional Details : CAS Number : 90604-29-8 Weight : 0.10000kg

 View more versions of this product

Product Code. 10056330

  • £299.00 / 100g

Estimated Shipment: 30-09-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Characteristics:
  • CAS number: 90604-29-8
  • EC number: 292-322-5
  • Bulk density: 150kg/m3
  • Solubility: 1g/L
  • Appearance: Slightly yellow, leafs
  • K (Potassium): ≤0.02%
  • Na (Sodium): ≤0.5%
  • Mg (Magnesium): ≤0.01%
  • Ca (Calcium): ≤0.05%
  • Heavy metals (as lead): ≤0.001%
  • Fats: ≤0.2%
  • Decomposition products: not detectable
  • Ash (600°C): ≤1%
  • NADH Oxidase / LDH pyruvate: not detectable
Specifications

Specifications

Albumin: ≥98.0%
Solid
6.8–7.2 (1g/L at 20°C)
Refrigerated (2°-8°C)
Glass bottle
Biochemistry
Bovine serum albumin
100g
≤5.00% (acc. to Karl Fischer)
Bovine serum
Videos
SDS
Documents
Certificates