missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine Plasma, Crystalline, 98%

For research use

Brand:  Alfa Aesar™ J64752.06

Code : NEW

Additional Details : CAS Number : 9048-46-8 Weight : 0.00500kg

 View more versions of this product

Product Code. 15406569

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

For lipid solubilization; also a blocking agent in Western blots
Lyophilized powder
5g
2° to 8°C
Albumin, Bovine Plasma, crystalline, 98%
Lyophilized Powder
98%
Bovine plasma
RUO
No
Videos
SDS

For Research Use Only