missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine, New Zealand origin, Standard Grade, Chromatographically Purified

For research use

Brand:  Alfa Aesar™ J64777.09

UNSPSC : 12352200

Code : NEW

Additional Details : CAS Number : 9048-46-8 Weight : 0.01000kg

 View more versions of this product

Product Code. 15456579

  • £23.11 / 10g


Estimated Shipment date
from Supplier 09-06-2017
Add to basket

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

Suitable for standard protein research applications
Lyophilized powder
Bovine Serum
RUO
No
Lyophilized Powder
10g
2° to 8°C
Albumin, Bovine, New Zealand origin, Standard grade, Chromatographically purified
Documentation

Technical Documents