Alfa Aesar™ Albumin, Bovine, New Zealand origin, RIA Grade, Chromatographically Purified

For research use

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J65189.18

50GR Albumin, Bovine, New Zealand origin, RIA Grade, Chromatographically purified

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: CAS Number: 9048-46-8 Weight: 0.05000kg



Disclaimers: For Research Use Only

Product Code. 15446779

Quantity Price
1 £ 221.96 / 50g
Estimated Shipment date
from Supplier 31-01-2017
Add to basket


Description and Specification

Specification

Source Bovine Serum
Quantity 50g
Recombinant No
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Albumin, Bovine, New Zealand origin, RIA grade, Chromatographically purified
For Use With (Application) For standard use in radioimmunoassays
Format Lyophilized powder
Formulation Lyophilized powder