missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine, New Zealand origin, Low Fatty Acid, Chromatographically Purified

For research use

Brand:  Alfa Aesar™ J65400.22

UNSPSC : 12352200

Code : NEW

Additional Details : CAS Number : 9048-46-8 Weight : 0.10000kg

 View more versions of this product

Product Code. 15446889

  • £171.30 / 100g


Estimated Shipment date
from Supplier 12-07-2017
Add to basket

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

Suitable for immunoassays. Reduced fatty acid content minimizes non-specific binding of antibodies
Lyophilized powder
Bovine Serum
RUO
No
Lyophilized Powder
100g
2° to 8°C
Albumin, Bovine, New Zealand origin, Low Fatty Acid, Chromatographically purified
Documentation

Technical Documents