Alfa Aesar™ Albumin, Bovine, New Zealand origin, Low Fatty Acid, Chromatographically Purified

For research use

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J65400.14

25GR Albumin, Bovine, New Zealand origin, Low Fatty Acid, Chromatographically purified

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: CAS Number: 9048-46-8 Weight: 0.02500kg



Disclaimers: For Research Use Only

Product Code. 15426889

Quantity Price
1 £ 65.51 / 25g
Estimated Shipment date
from Supplier 01-02-2017
Add to basket


Description and Specification

Specification

Quantity 25g
Recombinant No
Source Bovine Serum
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Albumin, Bovine, New Zealand origin, Low Fatty Acid, Chromatographically purified
For Use With (Application) Suitable for immunoassays. Reduced fatty acid content minimizes non-specific binding of antibodies
Format Lyophilized powder
Formulation Lyophilized powder