missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine, Fraction V, 98%, Reagent Grade, pH 7.0

For research use

Brand:  Alfa Aesar™ J64100.A1

UNSPSC : 12352200

Code : NEW

Additional Details : CAS Number : 9048-46-8 Weight : 1.00000kg

 View more versions of this product

Product Code. 15416209

  • POA / 1kg


Estimated Shipment date
from Supplier 10-07-2017
Add to basket

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

Lyophilized Powder
7.0
1kg
2° to 8°C
Albumin, Bovine, Fraction V, 98%, Reagent grade, pH 7.0
Lyophilized powder
Reagent grade, 98%
Bovine Serum
RUO
No
Documentation

Technical Documents