Alfa Aesar™ Albumin, Bovine, Fraction V, 98%, Reagent Grade, pH 7.0

For research use

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J64100.22

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: CAS Number: 9048-46-8 Weight: 0.10000kg



Disclaimers: For Research Use Only

Product Code. 15486199

Quantity Price
1 £ 154.01999999999998 / 100g
Estimated Shipment date
from Supplier 12-04-2017
Add to basket


Description and Specification

Specification

Quantity 100g
Recombinant No
Regulatory Status RUO
Formulation Lyophilized powder
Purity Reagent grade, 98%
Format Lyophilized powder
Storage Requirements 2° to 8°C
Product Type Albumin, Bovine, Fraction V, 98%, Reagent grade, pH 7.0
pH Range 7.0
Source Bovine Serum
Documentation

Technical documents