missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine, Fraction V, 97%, Standard Grade, pH 7.0

For research use

Brand:  Alfa Aesar™ J64655.22

Code : NEW

Additional Details : CAS Number : 9048-46-8 Weight : 0.10000kg

 View more versions of this product

Product Code. 15476499

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Lyophilized Powder
7.0
100g
2° to 8°C
Albumin, Bovine, Fraction V, 97%, Standard grade, pH 7.0
Lyophilized powder
Standard grade, 97%
Bovine Serum
RUO
No
Videos

For Research Use Only