missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine, Fraction V, 96%, Standard Grade, New Zealand origin, pH 7.0

For research use

Brand:  Alfa Aesar™ J65788.30

UNSPSC : 12352200

Code : NEW

Additional Details : CAS Number : 9048-46-8 Weight : 0.25000kg

 View more versions of this product

Product Code. 15437079

  • £245.60 / 250g


Estimated Shipment date
from Supplier 12-06-2017
Add to basket

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

Lyophilized Powder
7.0
250g
2° to 8°C
Albumin, Bovine, Fraction V, 96%, Standard grade, New Zealand origin, pH 7.0
Lyophilized powder
Standard grade, 96%
Bovine Serum
RUO
No
Documentation

Technical Documents