missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine, Fraction V, 35% Solution, Cell Culture Grade

For research use

Brand:  Alfa Aesar™ J64101.AE

UNSPSC : 12352200

Code : NEW

Additional Details : CAS Number : 9048-46-8 Weight : 0.00010kg

 View more versions of this product

Product Code. 15436209

  • £266.48 / 100mL


Estimated Shipment date
from Supplier 11-08-2017
Add to basket

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

35% w/v
Aqueous solution with 0.1% azide
Bovine Serum
RUO
No
Liquid
100mL
2° to 8°C
Albumin, Bovine, Fraction V, 35% soln., Cell Culture grade
Documentation

Technical Documents