Learn More
Alfa Aesar™ Albumin, Bovine, Fraction V, 35% Solution, Cell Culture Grade, No Preservative
For research use
Brand: Alfa Aesar™ J64248.AE
Code : NEW
Additional Details : CAS Number : 9048-46-8 Weight : 0.00010kg
Specifications
35% w/v | |
Aqueous solution | |
Bovine Serum | |
RUO | |
No |
Liquid | |
100mL | |
2° to 8°C | |
Albumin, Bovine, Fraction V, 35% soln., Cell Culture grade, no preservative |
For Research Use Only