missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine, Fraction V, 35% Solution, Cell Culture Grade

For research use

Brand:  Alfa Aesar™ J64101.AD

UNSPSC : 12352200

Code : NEW

Additional Details : CAS Number : 9048-46-8 Weight : 0.00050kg

 View more versions of this product

Product Code. 15426209

  • POA / 50mL


Estimated Shipment date
from Supplier 12-07-2017
Add to basket

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

35% w/v
Aqueous solution with 0.1% azide
Bovine Serum
RUO
No
Liquid
50mL
2° to 8°C
Albumin, Bovine, Fraction V, 35% soln., Cell Culture grade
Documentation

Technical Documents