missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine, Fraction V, 30% Solution, Standard Grade, No preservative

For research use

Brand:  Alfa Aesar™ J65833.AP

UNSPSC : 12352200

Code : NEW

Additional Details : CAS Number : 9048-46-8 Weight : 0.00050kg

 View more versions of this product

Product Code. 15467099

  • £329.24 / 500mL


Estimated Shipment date
from Supplier 08-06-2017
Add to basket

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

30% w/v
Liquid
Bovine Serum
RUO
No
Liquid
500mL
2° to 8°C
Albumin, Bovine, Fraction V, 30% soln., Standard grade, no preservative
Documentation

Technical Documents