missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine, Cohn Fraction V, 98%, Immunoassay Grade, Protease and Enzyme-free, pH5.2

For research use

Brand:  Alfa Aesar™ J65973.30

UNSPSC : 12352200

Code : NEW

Additional Details : CAS Number : 9048-46-8 Weight : 0.25000kg

 View more versions of this product

Product Code. 15487159

  • £326.27 / 250g


Estimated Shipment date
from Supplier 07-09-2017
Add to basket

Disclaimers:

For Research Use Only


Specifications

Specifications

Lyophilized Powder
5.2
250g
2° to 8°C
Albumin, Bovine, Cohn Fraction V, 98%, Immunoassay grade, protease and enzyme-free, pH5.2
Lyophilized powder
Immunoassay grade, 98%
Bovine Serum
RUO
No
Documentation

Technical Documents