Alfa Aesar™ Albumin, Bovine, Cohn Fraction V, 98%, Fatty Acid Free, pH 7.0

For research use

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J64944.A1

1KG Albumin, Bovine, Cohn Fraction V, 98%, Fatty Acid Free, pH 7.0

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: CAS Number: 9048-46-8 Weight: 1.00000kg



Disclaimers: For Research Use Only

Product Code. 15476659

Quantity Price
1 £ 864.01 / 1kg
Estimated Shipment date
from Supplier 02-02-2017
Add to basket


Description and Specification

Specification

pH Range 7.0
Purity 98%
Quantity 1kg
Recombinant No
Source Bovine Serum
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Albumin, Bovine, Cohn Fraction V, 98%, Fatty Acid Free, pH 7.0
Format Lyophilized powder
Formulation Lyophilized powder