missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine, Cohn Fraction V, 98%, Fatty Acid Free, pH 7.0

For research use

Brand:  Alfa Aesar™ J64944.36

Code : NEW

Additional Details : CAS Number : 9048-46-8 Weight : 0.50000kg

 View more versions of this product

Product Code. 15466659

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Lyophilized Powder
7.0
500g
2° to 8°C
Albumin, Bovine, Cohn Fraction V, 98%, Fatty Acid Free, pH 7.0
Lyophilized powder
98%
Bovine Serum
RUO
No
Videos
SDS

For Research Use Only