Alfa Aesar™ Albumin, Bovine, Cohn Fraction V, 98%, Cell Culture Grade, Low Endotoxin

For research use

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J65097.09

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: CAS Number: 9048-46-8 Weight: 0.01000kg



Disclaimers: For Research Use Only

Product Code. 15446719

Quantity Price
1 £ 24.34 / 10g
Estimated Shipment date
from Supplier 11-05-2017
Add to basket


Description and Specification

Specification

Formulation Lyophilized powder
Quantity 10g
Source Bovine Serum
Purity Cell Culture grade, 98%
Regulatory Status RUO
Product Type Albumin, Bovine, Cohn Fraction V, 98%, Cell Culture grade, Low Endotoxin
Storage Requirements 2° to 8°C
Format Lyophilized powder
Recombinant No
Documentation

Technical documents