missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Albumin, Bovine, Cohn Fraction V, 98%, Cell Culture Grade, Low Endotoxin

For research use

Brand:  Alfa Aesar™ J65097.22

Code : NEW

Additional Details : CAS Number : 9048-46-8 Weight : 0.10000kg

 View more versions of this product

Product Code. 15466719

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

Lyophilized Powder
Cell Culture grade, 98%
Bovine Serum
RUO
No
Lyophilized powder
100g
2° to 8°C
Albumin, Bovine, Cohn Fraction V, 98%, Cell Culture grade, Low Endotoxin
Videos
SDS

For Research Use Only