Learn More
alanyl-tRNA Synthetase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559335
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DRASKADVQK RVLEKTKQFI DSNPNQPLVI LEMESGASAK ALNEALKLFK MHSPQTSAML FTVDNEAGKI TCLCQVPQNA ANRGLKASEW VQQVSGLMDG KGGGKD Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
alanyl-tRNA Synthetase Polyclonal antibody specifically detects alanyl-tRNA Synthetase in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
alanyl-tRNA Synthetase | |
Polyclonal | |
Unconjugated | |
AARS1 | |
Aars; AI316495; alanine tRNA ligase 1, cytoplasmic; alanine--tRNA ligase, cytoplasmic; alanine--tRNA ligase, cytoplasmic {ECO:0000255; Alanyl-tRNA synthetase; alanyl-tRNA synthetase {ECO:0000255; alanyl-tRNA synthetase, cytoplasmic; alaRS; alaRS {ECO:0000255; C76919; CMT2N; EIEE29; HAMAP-Rule:MF_03133}; Protein sticky; Renal carcinoma antigen NY-REN-42; sti | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16, 234734, 292023 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49588, P50475, Q8BGQ7 | |
AARS1 | |
Recombinant protein corresponding to Human alanyl-tRNA Synthetase | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only