missing translation for 'onlineSavingsMsg'
Learn More

Alantolactone, Tocris Bioscience™

Antiproliferative; induces activin/Smad3 signaling

Brand:  RD Systems (Bio-Techne) 4490/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13464834

  • £609.76 / 50mg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sesquiterpene lactone. Induces activin/SMAD3 signaling and disrupts Cripto-1/activin receptor type II A interaction. Displays antiproliferative activity in human colon adenocarcinoma HCT-8 cells.
Specifications

Chemical Identifiers

546-43-0
232.323
alantolactone, helenine, eupatal, alant camphor, inula camphor, helenin, elecampane camphor, unii-m7gsn5q1m6, m7gsn5q1m6, 3ar,5s,8ar,9ar-5,8a-dimethyl-3-methylidene-3a,5,6,7,8,8a,9,9a-octahydronaphtho 2,3-b furan-2 3h-one
CHEBI:2540
CC1CCCC2(C1=CC3C(C2)OC(=O)C3=C)C
C15H20O2
PXOYOCNNSUAQNS-AGNJHWRGSA-N
72724
(3aR,5S,8aR,9aR)-5,8a-dimethyl-3-methylidene-5,6,7,8,9,9a-hexahydro-3aH-benzo[f][1]benzofuran-2-one

Specifications

Alantolactone
546-43-0
alantolactone, helenine, eupatal, alant camphor, inula camphor, helenin, elecampane camphor, unii-m7gsn5q1m6, m7gsn5q1m6, 3ar,5s,8ar,9ar-5,8a-dimethyl-3-methylidene-3a,5,6,7,8,8a,9,9a-octahydronaphtho 2,3-b furan-2 3h-one
CC1CCCC2(C1=CC3C(C2)OC(=O)C3=C)C
232.323
CHEBI:2540
>98%
50mg
C15H20O2
PXOYOCNNSUAQNS-AGNJHWRGSA-N
(3aR,5S,8aR,9aR)-5,8a-dimethyl-3-methylidene-5,6,7,8,9,9a-hexahydro-3aH-benzo[f][1]benzofuran-2-one
72724
232.32
Videos
Documents