missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ [Ala&superscript_2;]-Leu-Enkephalin

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J63207.MC

Code : NEW

Additional Details : CAS Number : 60284-47-1 Weight : 0.00010kg

 View more versions of this product

Product Code. 15435469

  • £501.17 / 100mg


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Specifications

569.66
100mg
Lyophilized powder
RUO
[Ala2]-Leu-Enkephalin
HPLC
An opioid peptide
Tyr-Ala-Gly-Phe-Leu
-30° to -10°C
Videos
SDS

For Research Use Only