missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar Al-25 Tube, Both ends open, O.D.: 70mm, I.D.: 54mm, Alfa Aesar™

Brand:  Alfa Aesar 031615.DM

Al-25 Tube, Both ends open, O.D.: 70mm, I.D.: 54mm, Alfa Aesar™

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11314167

  • £157.73 / 100mm


Estimated Shipment date
from Supplier 08-05-2018
Add to basket