missing translation for 'onlineSavingsMsg'
Learn More

Al-25 Tube, Both ends open, O.D.: 60mm, I.D.: 50mm, Alfa Aesar™

Brand:  Alfa Aesar 031614.ED

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 500mm

 View more versions of this product

Product Code. 11384157

  • £402.48 / 500mm


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

Al-25 Tube
Both ends open

Specifications

Al-25 Tube
500mm
Both ends open
Videos
SDS