missing translation for 'onlineSavingsMsg'
Learn More

Al-24 Tube, One End Closed, O.D.: 24mm, I.D.: 18mm, Alfa Aesar™

Brand:  Alfa Aesar 030093.DM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 100mm

 View more versions of this product

Product Code. 11322868

  • £160.10 / 100mm


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-24 Tube
One end closed

Specifications

Al-24 Tube
100mm
One end closed
Videos
SDS