missing translation for 'onlineSavingsMsg'
Learn More

Al-24 Tube, One End Closed, O.D.: 24mm, I.D.: 18mm, Alfa Aesar™

Brand:  Alfa Aesar 030093.EM

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 800mm

 View more versions of this product

Product Code. 11312868

  • £291.10 / 800mm


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

Al-24 Tube
One end closed

Specifications

Al-24 Tube
800mm
One end closed
Videos
SDS