missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Al-24 Tube, One End Closed, O.D.: 15mm, I.D.: 10mm, Alfa Aesar™

Brand:  Thermo Scientific Alfa Aesar 030091.DW

Al-24 Tube, One End Closed, O.D.: 15mm, I.D.: 10mm, Alfa Aesar™

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11342858

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.