missing translation for 'onlineSavingsMsg'
Learn More

Al-24 Tube, One End Closed, O.D.: 15mm, I.D.: 10mm, Alfa Aesar™

Brand:  Alfa Aesar 030091.EO

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1000mm

 View more versions of this product

Product Code. 11382858

  • £124.87 / 1000mm


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

Al-24 Tube
One end closed

Specifications

Al-24 Tube
1000mm
One end closed
Videos
SDS