missing translation for 'onlineSavingsMsg'
Learn More

Al-24 Tube, One End Closed, O.D.: 15mm, I.D.: 10mm, Alfa Aesar™

Brand:  Alfa Aesar 030091.EM

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11302868

  • £106.21 / 800mm


Estimated Shipment date
from Supplier 05-07-2018
Add to basket

Specifications

Specifications

Al-24 Tube
800mm
One end closed
Videos
SDS