missing translation for 'onlineSavingsMsg'
Learn More

Al-24 Tube, Both ends open, O.D.: 70mm, I.D.: 54mm, Alfa Aesar™

Brand:  Alfa Aesar 031610.EO

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11384137

  • £1283.34 / 1000mm


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Specifications

Al-24 Tube
1000mm
Both ends open
Videos
SDS