missing translation for 'onlineSavingsMsg'
Learn More

Al-24 Tube, Both ends open, O.D.: 40mm, I.D.: 32mm, Alfa Aesar™

Brand:  Alfa Aesar 031607.EH

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11364117

  • £343.44 / 600mm


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Specifications

Al-24 Tube
600mm
Both ends open
Videos
SDS