missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Al-24 Tube, Both ends open, O.D.: 40mm, I.D.: 32mm, Alfa Aesar™

Brand:  Thermo Scientific Alfa Aesar 031607.DM

Al-24 Tube, Both ends open, O.D.: 40mm, I.D.: 32mm, Alfa Aesar™

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11394117

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.