missing translation for 'onlineSavingsMsg'
Learn More

Al-24 Tube, Both ends open, O.D.: 30mm, I.D.: 24mm, Alfa Aesar™

Brand:  Alfa Aesar 031606.EO

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1000mm

 View more versions of this product

Product Code. 11314117

  • £310.40 / 1000mm


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-24 Tube
Both ends open

Specifications

Al-24 Tube
1000mm
Both ends open
Videos
SDS