missing translation for 'onlineSavingsMsg'
Learn More

Al-24 Tube, Both ends open, O.D.: 20mm, I.D.: 15mm, Alfa Aesar™

Brand:  Alfa Aesar 031604.EH

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 600mm

 View more versions of this product

Product Code. 11374097

  • £129.74 / 600mm


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Al-24 Tube
Both ends open

Specifications

Al-24 Tube
600mm
Both ends open
Videos
SDS